Diethyl (3-chlorobenzyl)phosphonate ≥98.0%

Supplier: TCI America

C1595-5G C1595-1G
TCC1595-5GEA 69.52 CAD
TCC1595-5G TCC1595-001G
Diethyl (3-chlorobenzyl)phosphonate ≥98.0%
Diethyl (3-chlorobenzyl)phosphonate

CAS Number: 78055-64-8
MDL Number: MFCD01861282
Molecular Formula: C11H16ClO3P
Molecular Weight: 262.67
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 163
Flash Point (°C): 110
Specific Gravity (20/20): 1.19


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (3-Chlorobenzyl)phosphonic Acid Diethyl Ester

Formula: C₁₁H₁₆ClO₃P
Boiling Pt: 163 °C
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01861282
CAS Number: 78055-64-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51eba04e1b8311361c61e116d08561c69ce22eba&vid=51eba04e1b8311361c61e116d08561c69ce22eba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR