1-Chloro-2,5-diethoxybenzene ≥98.0%
Supplier: TCI America
C1576-25G
C1576-500G
TCC1576-025GEA
45.82
CAD
TCC1576-025G
TCC1576-500G
1-Chloro-2,5-diethoxybenzene ≥98.0%
1-Chloro-2,5-diethoxybenzene
CAS Number: 52196-74-4
MDL Number: MFCD00026765
Molecular Formula: C10H13ClO2
Molecular Weight: 200.66
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 266
Flash Point (°C): 130
Specific Gravity (20/20): 1.14
Other Product Names / Synonyms: 2,5-Diethoxychlorobenzene
Formula:
C₁₀H₁₃ClO₂ Boiling Pt: 266 °C Density: 1.14 (20 °C) Flash Pt: 130 °C |
MDL Number:
MFCD00026765 CAS Number: 52196-74-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...