Chloromethyl phenyl sulfone ≥98.0%

Supplier: TCI America

Synonyms: ((Chloromethyl)sulphonyl)benzene, 1-(Chloromethylsulfonyl)benzene, 1-(Chloromethylsulphonyl)benzene, Phenyl Chloromethyl Sulphone, Phenyl Chloromethyl sulfone, ((Chloromethyl)sulfonyl)benzene

C1571-25G C1571-5G
TCC1571-025GEA 698.36 CAD
TCC1571-025G TCC1571-005G
Chloromethyl phenyl sulfone ≥98.0%
Chloromethyl phenyl sulfone

CAS Number: 7205-98-3
MDL Number: MFCD00007551
Molecular Formula: C7H7ClO2S
Molecular Weight: 190.64
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 53
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Nucleophiles (Aromatic VNS Reaction)
Other Product Names / Synonyms: Phenyl Chloromethyl Sulfone

Formula: C₇H₇ClO₂S
Boiling Pt: 130 °C
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD00007551
CAS Number: 7205-98-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d174bfbba2c072455579df5d795cf4b04912d813&vid=d174bfbba2c072455579df5d795cf4b04912d813&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR