2-Chloro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C1560-1G C1560-100MG
TCC1560-1GEA 1053.86 CAD
TCC1560-1G TCC1560-100MG
2-Chloro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
2-Chloro-1,10-phenanthroline

CAS Number: 7089-68-1
MDL Number: MFCD00185678
Molecular Formula: C12H7ClN2
Molecular Weight: 214.65
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130

Formula: C₁₂H₇ClN₂
MW: 214.65 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00185678
CAS Number: 7089-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d33ddcc1676cff86ddf6d14e82b11f538752e6f0&vid=d33ddcc1676cff86ddf6d14e82b11f538752e6f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR