4-Chloropyridine-N-oxide ≥97.0%

Supplier: TCI America

C1540-5G C1540-25G
TCC1540-005GEA 278.08 CAD
TCC1540-005G TCC1540-25G
4-Chloropyridine-N-oxide ≥97.0%
4-Chloropyridine N-oxide

CAS Number: 1121-76-2
MDL Number: MFCD00047425
Molecular Formula: C5H4ClNO
Molecular Weight: 129.54
Purity/Analysis Method: >97.0% (GC)
Form: Crystal

Formula: C₅H₄ClNO
MDL Number: MFCD00047425
CAS Number: 1121-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c496f0d472807fd296374a4505e70a90536f6ee0&vid=c496f0d472807fd296374a4505e70a90536f6ee0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR