3-Chloro-2,2-dimethyl-1-propanol ≥98.0%
Supplier: TCI America
C1538-25G
C1538-500G
TCC1538-025GEA
60.04
CAD
TCC1538-025G
TCC1538-500G
3-Chloro-2,2-dimethyl-1-propanol ≥98.0%
3-Chloro-2,2-dimethyl-1-propanol
CAS Number: 13401-56-4
MDL Number: MFCD00004684
Molecular Formula: C5H11ClO
Molecular Weight: 122.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 71
Freezing point (°C): 35
Formula:
C₅H₁₁ClO Flash Pt: 71 °C |
MDL Number:
MFCD00004684 CAS Number: 13401-56-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...