N-Carbobenzoxy-1,5-diaminopentane hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1520-5G C1520-1G
TCC1520-005GEA 623.15 CAD
TCC1520-005G TCC1520-001G
N-Carbobenzoxy-1,5-diaminopentane hydrochloride ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-1,5-diaminopentane hydrochloride

CAS Number: 18807-74-4
MDL Number: MFCD00270152
Molecular Formula: C13H20N2O2
Molecular Weight: 272.77
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 172


Product Categories: Bioscience, Chemical Building Blocks for Polyamines

Formula: C₁₃H₂₀N₂O₂
Melting Pt: 172 °C
MDL Number: MFCD00270152
CAS Number: 18807-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84a73bd4abc15fc3a280fea67b7ea256be3280c7&vid=84a73bd4abc15fc3a280fea67b7ea256be3280c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR