N-Carbobenzoxy-1,4-diaminobutane hydrochloride ≥97.0%
Supplier: TCI America
Synonyms:
N-Z-1,4-Butanediamine hydrochloride, N-Cbz-1,4-diaminobutane Hydrochloride, Benzyl N-(4-Aminobutyl)carbamate Hydrochloride, Benzyl (4-aminobutyl)carbamate hydrochloride, N-(4-Aminobutyl)carbamic Acid Benzyl Ester Hydrochloride
CAS Number: 18807-73-3
MDL Number: MFCD00270149
Molecular Formula: C12H18N2O2
Molecular Weight: 258.75
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-(4-Aminobutyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(4-Aminobutyl)carbamate Hydrochloride, N-Cbz-1,4-diaminobutane Hydrochloride
Formula:
C₁₂H₁₈N₂O₂ MW: 258.75 g/mol |
MDL Number:
MFCD00270149 CAS Number: 18807-73-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...