N-Carbobenzoxy-1,3-diaminopropane hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C1512-1G
C1512-5G
TCC1512-001GEA
50.56
CAD
TCC1512-001G
TCC1512-005G
N-Carbobenzoxy-1,3-diaminopropane hydrochloride ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-1,3-diaminopropane hydrochloride
CAS Number: 17400-34-9
MDL Number: MFCD00270153
Molecular Formula: C11H16N2O2
Molecular Weight: 244.72
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 186
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-(3-Aminopropyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(3-Aminopropyl)carbamate Hydrochloride, N-Cbz-1,3-diaminopropane Hydrochloride
Formula:
C₁₁H₁₆N₂O₂ Melting Pt: 186 °C |
MDL Number:
MFCD00270153 CAS Number: 17400-34-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...