Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1511-25G C1511-1G C1511-5G
TCC1511-25GEA 472.42 CAD
TCC1511-25G TCC1511-001G TCC1511-005G
Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥98.0% (by titrimetric analysis)
Benzyl N-(2-aminoethyl)carbamate hydrochloride

CAS Number: 18807-71-1
MDL Number: MFCD00270150
Molecular Formula: C10H14N2O2
Molecular Weight: 230.69
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 172


Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-(2-Aminoethyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(2-Aminoethyl)carbamate Hydrochloride, N-Cbz-1,2-diaminoethane Hydrochloride

Formula: C₁₀H₁₄N₂O₂
Melting Pt: 172 °C
MDL Number: MFCD00270150
CAS Number: 18807-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc6c7a3a20a1203d753c3710d25a8b4d2aec3f43&vid=bc6c7a3a20a1203d753c3710d25a8b4d2aec3f43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR