3-(Hydroxymethyl)benzonitrile ≥98.0%

Supplier: TCI America

C1510-1G C1510-5G
TCC1510-001GEA 50.56 CAD
TCC1510-001G TCC1510-005G
3-(Hydroxymethyl)benzonitrile ≥98.0%
3-(Hydroxymethyl)benzonitrile

CAS Number: 874-97-5
MDL Number: MFCD01321087
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 109
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 3-(Hydroxymethyl)benzonitrile

Formula: C₈H₇NO
Boiling Pt: 165 °C
Density: 1.13 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD01321087
CAS Number: 874-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=602d0d7d0e62243649ecdb932acf1bcceaf0364b&vid=602d0d7d0e62243649ecdb932acf1bcceaf0364b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR