3-Carbamoylpicolinic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

C1508-1G C1508-100MG
TCC1508-1GEA 1005.04 CAD
TCC1508-1G TCC1508-100MG
3-Carbamoylpicolinic acid ≥97.0% (by titrimetric analysis)
3-Carbamoylpicolinic acid

CAS Number: 4733-65-7
MDL Number: MFCD00661289
Molecular Formula: C7H6N2O3
Molecular Weight: 166.14
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: 3-Carbamoylpicolinic Acid, 2-Carboxypyridine-3-carboxamide

Formula: C₇H₆N₂O₃
MDL Number: MFCD00661289
CAS Number: 4733-65-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2495918ca134e6a37f77833ab0e5f071d7f05f75&vid=2495918ca134e6a37f77833ab0e5f071d7f05f75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR