Cyanomethylenetributylphosphorane ≥95.0% (by titrimetric analysis)

Supplier: TCI America

C1500-5G C1500-25G C1500-1G
TCC1500-5GEA 930.4 CAD
TCC1500-5G TCC1500-25G TCC1500-001G
Cyanomethylenetributylphosphorane ≥95.0% (by titrimetric analysis)
Cyanomethylenetributylphosphorane

CAS Number: 157141-27-0
MDL Number: MFCD01321151
Molecular Formula: C14H28NP
Molecular Weight: 241.36
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C

Formula: C₁₄H₂₈NP
Density: 0.92 (20 °C)
MDL Number: MFCD01321151
CAS Number: 157141-27-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cacdb69190afdf57137ac8f54b2fb893f5bffdee&vid=cacdb69190afdf57137ac8f54b2fb893f5bffdee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR