Isoamyl acetoacetate ≥97.0%

Supplier: TCI America

Synonyms: Isopentyl Acetoacetate, Acetoacetic Acid Isoamyl Ester

A0812-500ML A0812-25ML
TCA0812-500MLEA 827.92 CAD
TCA0812-500ML TCA0812-025ML
Isoamyl acetoacetate ≥97.0%
Isoamyl acetoacetate

CAS Number: 2308-18-1
MDL Number: MFCD00059355
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 218
Flash Point (°C): 101
Specific Gravity (20/20): 0.97


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Acetoacetic Acid Isoamyl Ester, Isopentyl Acetoacetate

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
Boiling Pt: 218 °C
Density: 0.97 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00059355
CAS Number: 2308-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ced3132ee7404dac8085854b611d06ff1a281229&vid=ced3132ee7404dac8085854b611d06ff1a281229&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR