N-(Cyclohexylthio)phthalimide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

C1452-500G C1452-25G
TCC1452-500GEA 252.8 CAD
TCC1452-500G TCC1452-025G
N-(Cyclohexylthio)phthalimide ≥98.0% (by total nitrogen basis)
N-(Cyclohexylthio)phthalimide

CAS Number: 17796-82-6
MDL Number: MFCD00072247
Molecular Formula: C14H15NO2S
Molecular Weight: 261.34
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 91
Flash Point (°C): 270


Product Categories: Synthetic Organic Chemistry, Sulfenylation
Other Product Names / Synonyms: Cyclohexyl N-Phthalimidyl Sulfide

Formula: C₁₄H₁₅NO₂S
Melting Pt: 91 °C
Flash Pt: 270 °C
MDL Number: MFCD00072247
CAS Number: 17796-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9b25f590e75d39e46baaf3ecf47e60ecf2b3a10&vid=a9b25f590e75d39e46baaf3ecf47e60ecf2b3a10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR