(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (1S)-trans-1,2-Diaminocyclohexane D-Tartrate, (1S,2S)-Cyclohexane-1,2-diamine (2S,3S)-2,3-dihydroxysuccinate

C1450-5G C1450-25G
TCC1450-005GEA 49.61 CAD
TCC1450-005G TCC1450-025G
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate ≥98.0% (by titrimetric analysis)
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate

CAS Number: 67333-70-4
MDL Number: MFCD00191980
Molecular Formula: C6H14N2
Molecular Weight: 264.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 284
Specific rotation [a]20/D: -13 deg (C=2.5, H2O)


Other Product Names / Synonyms: (1S)-trans-1,2-Diaminocyclohexane D-Tartrate

Formula: C₆H₁₄N₂
MW: 264.28 g/mol
Melting Pt: 284 °C
MDL Number: MFCD00191980
CAS Number: 67333-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79e894a885ad3df475dab7b3e4010f7aeb39b22e&vid=79e894a885ad3df475dab7b3e4010f7aeb39b22e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR