N-(Carbobenzoxy)aminoacetonitrile ≥95.0%

Supplier: TCI America

Synonyms: Benzyl (cyanomethyl)carbamate, Cyanomethylcarbamic Acid Benzyl Ester, Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile

C1439-25G C1439-5G
TCC1439-25GEA 1368.28 CAD
TCC1439-25G TCC1439-005G
N-(Carbobenzoxy)aminoacetonitrile ≥95.0%
N-(Carbobenzoxy)aminoacetonitrile

CAS Number: 3589-41-1
MDL Number: MFCD00033175
Molecular Formula: C10H10N2O2
Molecular Weight: 190.20
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Melting point (°C): 59


Other Product Names / Synonyms: Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile, Cyanomethylcarbamic Acid Benzyl Ester

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
Melting Pt: 59 °C
MDL Number: MFCD00033175
CAS Number: 3589-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abe18278637891bfa02a4ec3d2ac8dfbbda3189b&vid=abe18278637891bfa02a4ec3d2ac8dfbbda3189b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR