Diglycidyl-1,2-cyclohexanedicarboxylate (so called)

Supplier: TCI America

C1434-25G C1434-500G
TCC1434-025GEA 62.68 CAD
TCC1434-025G TCC1434-500G
Diglycidyl-1,2-cyclohexanedicarboxylate (so called)
Diglycidyl-1,2-cyclohexanedicarboxylate

CAS Number: 5493-45-8
MDL Number: MFCD00128914
Molecular Formula: C14H20O6
Molecular Weight: 284.31
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 205
Flash Point (°C): 113
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: 1,2-Cyclohexanedicarboxylic Acid Diglycidyl Ester

Formula: C₁₄H₂₀O₆
Boiling Pt: 205 °C
Density: 1.22 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00128914
CAS Number: 5493-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37c45ac449d4ff581e1f93f7a9ae9bb5ef84411b&vid=37c45ac449d4ff581e1f93f7a9ae9bb5ef84411b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR