(Chloromethyl)dimethylphenylsilane ≥97.0%

Supplier: TCI America

C1419-25G C1419-5G
TCC1419-25GEA 396.58 CAD
TCC1419-25G TCC1419-5G
(Chloromethyl)dimethylphenylsilane ≥97.0%
(Chloromethyl)dimethylphenylsilane

CAS Number: 1833-51-8
MDL Number: MFCD00040858
Molecular Formula: C9H13ClSi
Molecular Weight: 184.74
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 91
Specific Gravity (20/20): 1.03

Formula: C₉H₁₃ClSi
Boiling Pt: 225 °C
Density: 1.03 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00040858
CAS Number: 1833-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55e6d3488da37ad3b4b7ea207916893830c76ffb&vid=55e6d3488da37ad3b4b7ea207916893830c76ffb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR