(-)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0%

Supplier: TCI America

C1418-100MG C1418-1G
TCC1418-100MGEA 135.56 CAD
TCC1418-100MG TCC1418-1G
(-)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0%
(-)-trans-1,2-Cyclohexanedicarboxylic anhydride

CAS Number: 31982-85-1
MDL Number: MFCD00674195
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 166
Specific rotation [a]20/D: -85 deg (C=1, Dioxane)

Formula: C₈H₁₀O₃
Melting Pt: 166 °C
MDL Number: MFCD00674195
CAS Number: 31982-85-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d38c964b854249122c308b0b62603754cb701abf&vid=d38c964b854249122c308b0b62603754cb701abf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR