(+)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0%
Supplier: TCI America
Synonyms:
(3aR,7aR)-Hexahydroisobenzofuran-1,3-dione
C1417-100MG
C1417-1G
TCC1417-100MGEA
158.63
CAD
TCC1417-100MG
TCC1417-1G
(+)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0%
(+)-trans-1,2-Cyclohexanedicarboxylic anhydride
CAS Number: 71749-03-6
MDL Number: MFCD00674194
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 165
Specific rotation [a]20/D: 85 deg (C=1, Dioxane)
Formula:
C₈H₁₀O₃ Melting Pt: 165 °C |
MDL Number:
MFCD00674194 CAS Number: 71749-03-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...