2-Chloro-1,3-dimethylimidazolinium chloride ≥98.0% (by HPLC)

Supplier: TCI America

C1408-5G C1408-25G
TCC1408-005GEA 92.59 CAD
TCC1408-005G TCC1408-025G
2-Chloro-1,3-dimethylimidazolinium chloride ≥98.0% (by HPLC)
2-Chloro-1,3-dimethylimidazolinium chloride

CAS Number: 37091-73-9
MDL Number: MFCD00137463
Molecular Formula: C5H10Cl2N2
Molecular Weight: 169.05
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: DMC

Formula: C₅H₁₀Cl₂N₂
MDL Number: MFCD00137463
CAS Number: 37091-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fff0ca62971c3e451cd2a0889d26769bb59949c2&vid=fff0ca62971c3e451cd2a0889d26769bb59949c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR