Dipyridin-2-ylcarbonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C1407-5G
C1407-1G
TCC1407-005GEA
205.4
CAD
TCC1407-005G
TCC1407-1G
Dipyridin-2-ylcarbonate ≥98.0% (by titrimetric analysis)
Dipyridin-2-ylcarbonate
CAS Number: 1659-31-0
MDL Number: MFCD00191407
Molecular Formula: C11H8N2O3
Molecular Weight: 216.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 90
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: Carbonic Acid Di-2-pyridyl Ester
Formula:
C₁₁H₈N₂O₃ Melting Pt: 90 °C |
MDL Number:
MFCD00191407 CAS Number: 1659-31-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...