(±)-2-Amino-2-(4-chlorophenyl)acetic acid 98,0

Supplier: TCI America

Synonyms: DL-2-(4-Chlorophenyl)glycine, H-DL-Phg(4-Cl)-OH, DL-alpha-Amino-4-chlorophenylacetic acid, 4-Chloro-DL-2-phenylglycine, 4-Chloro-DL-phenylglycine, 2-Amino-2-(4-chlorophenyl)acetic acid

C1401-5G C1401-1G C1401-25G
TCC1401-5GEA 34.76 CAD
TCC1401-5G TCC1401-001G TCC1401-25G
(±)-2-Amino-2-(4-chlorophenyl)acetic acid 98,0
(±)-2-Amino-2-(4-chlorophenyl)acetic acid

CAS Number: 6212-33-5
MDL Number: MFCD00049328
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-alpha-Amino-4-chlorophenylacetic Acid, 4-Chloro-DL-2-phenylglycine, H-DL-Phg(4-Cl)-OH

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD00049328
CAS Number: 6212-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f16eb39531f543ee42b24468ee5f350aaa694968&vid=f16eb39531f543ee42b24468ee5f350aaa694968&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR