2-Ethylhexyl cyanoacetate ≥98.0%

Supplier: TCI America

Synonyms: Octyl Cyanoacetate, Cyanoacetic Acid 2-Ethylhexyl Ester, 2-Ethylhexyl Cyanoacetate, EHCA, Cyanoacetic Acid Octyl Ester

C1384-25ML C1384-500ML
TCC1384-025MLEA 26.86 CAD
TCC1384-025ML TCC1384-500ML
2-Ethylhexyl cyanoacetate ≥98.0%
2-Ethylhexyl cyanoacetate

CAS Number: 13361-34-7
MDL Number: MFCD00019843
Molecular Formula: C11H19NO2
Molecular Weight: 197.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 94
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Cyanoacetic Acid Octyl Ester, EHCA, Cyanoacetic Acid 2-Ethylhexyl Ester, Octyl Cyanoacetate

Formula: C₁₁H₁₉NO₂
MW: 197.28 g/mol
Density: 0.95 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00019843
CAS Number: 13361-34-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54f5d4cd4d490719302f81011af3df4d6e6e5c70&vid=54f5d4cd4d490719302f81011af3df4d6e6e5c70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR