N-Carbobenzoxy-O-tert-butyl-L-serine ≥98.0%

Supplier: TCI America

Synonyms: Z-Ser(tBu)-OH, N-Cbz-O-tert-butyl-L-serine, (S)-2-(((Benzyloxy)carbonyl)amino)-3-(tert-butoxy)propanoic acid, Cbz-Ser-OtBu

C1381-5G C1381-1G
TCC1381-005GEA 151.68 CAD
TCC1381-005G TCC1381-001G
N-Carbobenzoxy-O-tert-butyl-L-serine ≥98.0%
N-Carbobenzoxy-O-tert-butyl-L-serine

CAS Number: 1676-75-1
MDL Number: MFCD00038275
Molecular Formula: C15H21NO5
Molecular Weight: 295.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 86
Specific rotation [a]20/D: 21 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-O-tert-butyl-L-serine, Z-Ser(tBu)-OH

Formula: C₁₅H₂₁NO₅
MW: 295.34 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00038275
CAS Number: 1676-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17fa534472ff0e51fd303d068baffcb8ec86e740&vid=17fa534472ff0e51fd303d068baffcb8ec86e740&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR