1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-(Chloro(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium hexafluorophosphate(V), Chloro-N,N,N',N'-bis(tetramethylene)formamidinium Hexafluorophosphate, PyClU

C1379-5G C1379-25G
TCC1379-005GEA 75.84 CAD
TCC1379-005G TCC1379-25G
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate ≥97.0% (by titrimetric analysis)
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate

CAS Number: 135540-11-3
MDL Number: MFCD00191333
Molecular Formula: C9H16ClF6N2P
Molecular Weight: 332.66
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 154


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: Chloro-N,N,N',N'-bis(tetramethylene)formamidinium Hexafluorophosphate, PyClU

Formula: C₉H₁₆ClF₆N₂P
MW: 332.66 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00191333
CAS Number: 135540-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14a074b77045e1774ac9e1cc517c3cc56e7eb0f2&vid=14a074b77045e1774ac9e1cc517c3cc56e7eb0f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR