(2S,3S)-2-Chloro-3-methylvaleric acid ≥95.0%

Supplier: TCI America

Synonyms: (2S,3S)-2-Chloro-3-methylpentanoic Acid

C1372-1G C1372-5G
TCC1372-001GEA 153.73 CAD
TCC1372-001G TCC1372-005G
(2S,3S)-2-Chloro-3-methylvaleric acid ≥95.0%
(2S,3S)-2-Chloro-3-methylvaleric acid

CAS Number: 32653-34-2
MDL Number: MFCD00191403
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: -4.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (2S,3S)-2-Chloro-3-methylpentanoic Acid

Formula: C₆H₁₁ClO₂
MW: 150.60 g/mol
Boiling Pt: 125 °C
Density: 1.12 (20 °C)
MDL Number: MFCD00191403
CAS Number: 32653-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=beca2b7bf1ea1e8adc1e654fce2596afbb47b49b&vid=beca2b7bf1ea1e8adc1e654fce2596afbb47b49b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR