(S)-2-Chloro-4-methylvaleric acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

C1371-5G C1371-1G
TCC1371-005GEA 388.68 CAD
TCC1371-005G TCC1371-001G
(S)-2-Chloro-4-methylvaleric acid ≥97.0% (by titrimetric analysis)
(S)-2-Chloro-4-methylvaleric acid

CAS Number: 28659-81-6
MDL Number: MFCD00044943
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 126
Specific Gravity (20/20): 1.09
Specific rotation [a]20/D: -34 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (S)-2-Chloro-4-methylpentanoic Acid

Formula: C₆H₁₁ClO₂
Boiling Pt: 126 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00044943
CAS Number: 28659-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83621cae9df4a7ccda1b55bcb294cba344eb3841&vid=83621cae9df4a7ccda1b55bcb294cba344eb3841&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR