Methyl acetylsalicylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-acetoxybenzoate, Methyl 2-Acetoxybenzoate, Aspirin methyl ester, Acetylsalicylic Acid Methyl Ester, 2-Acetoxybenzoic Acid Methyl Ester

A0114-25G A0114-500G
TCA0114-025GEA 49.61 CAD
TCA0114-025G TCA0114-500G
Methyl acetylsalicylate ≥98.0%
Methyl acetylsalicylate

CAS Number: 580-02-9
MDL Number: MFCD00014978
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 50


Other Product Names / Synonyms: 2-Acetoxybenzoic Acid Methyl Ester, Methyl 2-Acetoxybenzoate, Acetylsalicylic Acid Methyl Ester

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 136 °C
Melting Pt: 50 °C
MDL Number: MFCD00014978
CAS Number: 580-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a427324d4668c9f7e85b605dc935cb05cd147c1&vid=7a427324d4668c9f7e85b605dc935cb05cd147c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR