1-Chloro-2,3-dimethoxybenzene ≥97.0%

Supplier: TCI America

C1355-5G C1355-1G
TCC1355-5GEA 537.2 CAD
TCC1355-5G TCC1355-001G
1-Chloro-2,3-dimethoxybenzene ≥97.0%
1-Chloro-2,3-dimethoxybenzene

CAS Number: 90282-99-8
MDL Number: MFCD00191399
Molecular Formula: C8H9ClO2
Molecular Weight: 172.61
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Specific Gravity (20/20): 1.21

Formula: C₈H₉ClO₂
Boiling Pt: 98 °C
Density: 1.21 (20 °C)
MDL Number: MFCD00191399
CAS Number: 90282-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72c8deafaf1cb990a2b9c872e4cb1e73d11af2bd&vid=72c8deafaf1cb990a2b9c872e4cb1e73d11af2bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR