4-Chloro-2,1,3-benzoxadiazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C1337-5G
C1337-25G
TCC1337-005GEA
266.8
CAD
TCC1337-005G
TCC1337-025G
4-Chloro-2,1,3-benzoxadiazole ≥98.0% (by titrimetric analysis)
4-Chloro-2,1,3-benzoxadiazole
CAS Number: 7116-16-7
MDL Number: MFCD00191329
Molecular Formula: C6H3ClN2O
Molecular Weight: 154.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 82
Other Product Names / Synonyms: 4-Chlorobenzofurazan
Formula:
C₆H₃ClN₂O Melting Pt: 82 °C |
MDL Number:
MFCD00191329 CAS Number: 7116-16-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...