D-Cysteine hydrochloride monohydrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

C1329-25G C1329-5G
TCC1329-25GEA 515.08 CAD
TCC1329-25G TCC1329-005G
D-Cysteine hydrochloride monohydrate ≥99.0% (by titrimetric analysis)
D-Cysteine hydrochloride monohydrate

CAS Number: 207121-46-8
MDL Number: MFCD00150051
Molecular Formula: C3H7NO2S
Molecular Weight: 157.61
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Specific rotation [a]20/D: -6 deg (C=8, 1mol/L HCl)


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Cys-OH.HCl

Formula: C₃H₇NO₂S
MDL Number: MFCD00150051
CAS Number: 207121-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdf7f1a065eff3615aecc049f1f315d9735df7a7&vid=cdf7f1a065eff3615aecc049f1f315d9735df7a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR