(2S,8aR)-(-)-(Camphorylsulfonyl)oxaziridine 95,0 Titrimetric

Supplier: TCI America

Synonyms: (1R,2S)-(-)-2,N-Epoxy-10,2-camphorsultam

C1327-1G C1327-5G
TCC1327-001GEA 44.24 CAD
TCC1327-001G TCC1327-5G
(2S,8aR)-(-)-(Camphorylsulfonyl)oxaziridine 95,0 Titrimetric
(2S,8aR)-(-)-(Camphorylsulfonyl)oxaziridine

[Asymmetric Oxidizing Reagent]
CAS Number: 104372-31-8
MDL Number: MFCD00075428
Molecular Formula: C10H15NO3S
Molecular Weight: 229.29
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 168
Specific rotation [a]20/D: -50 deg (C=1, Acetone)

Formula: C₁₀H₁₅NO₃S
MW: 229.29 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00075428
CAS Number: 104372-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a0a173c78d2cfb760217959950e292dbe5a0a67&vid=e7e2a9b5336d347c218f39bc44b39e8b49f31495&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR