(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide ≥95.0%

Supplier: TCI America

Synonyms: (1S)-(+)-(10-Camphorsulfonyl)oxaziridine, (4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide, (+)-(2R,8aS)-(Camphorylsulphonyl)oxaziridine, (1S)-(+)-(10-Camphorsulphonyl)oxaziridine, (+)-(2R,8aS)-(Camphorylsulfonyl)oxaziridine

C1326-1G C1326-5G
TCC1326-001GEA 74.26 CAD
TCC1326-001G TCC1326-5G
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide ≥95.0%
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide

[Asymmetric Oxidizing Reagent]
CAS Number: 104322-63-6
MDL Number: MFCD00075319
Molecular Formula: C10H15NO3S
Molecular Weight: 229.29
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: 50 deg (C=1, Acetone)

Formula: C₁₀H₁₅NO₃S
MW: 229.29 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00075319
CAS Number: 104322-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4a898ce89f038987fbb87010c4b3dc56f9b7756&vid=c4a898ce89f038987fbb87010c4b3dc56f9b7756&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR