[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

C1323-5G C1323-1G
TCC1323-5GEA 682.56 CAD
TCC1323-5G TCC1323-001G
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid ≥99.0% (by HPLC, titration analysis)
[4-Chloro-6-(2,3-xylidino)-2-pyrimidinylthio]acetic acid

CAS Number: 50892-23-4
MDL Number: MFCD00191335
Molecular Formula: C14H14ClN3O2S
Molecular Weight: 323.80
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 155

Formula: C₁₄H₁₄ClN₃O₂S
Melting Pt: 155 °C
MDL Number: MFCD00191335
CAS Number: 50892-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcc86de3405d5b3d60696420ee2f4e91ddd69844&vid=fcc86de3405d5b3d60696420ee2f4e91ddd69844&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR