Methyl-4-chloro-2-methylbutyrate ≥98.0%
Supplier: TCI America
C1319-25G
C1319-5G
TCC1319-025GEA
263.86
CAD
TCC1319-025G
TCC1319-5G
Methyl-4-chloro-2-methylbutyrate ≥98.0%
Methyl-4-chloro-2-methylbutyrate
CAS Number: 13888-03-4
MDL Number: MFCD00198001
Molecular Formula: C6H11ClO2
Molecular Weight: 150.60
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: 4-Chloro-2-methylbutyric Acid Methyl Ester
Formula:
C₆H₁₁ClO₂ Boiling Pt: 81 °C Density: 1.08 (20 °C) |
MDL Number:
MFCD00198001 CAS Number: 13888-03-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...