Ethyl-2,4-dihydroxy-6-methylbenzoate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ethyl 2,4-dihydroxy-6-methylbenzoate, 4-Ethoxycarbonyl-5-methylresorcinol, 2-Ethoxycarbonyl-3,5-dihydroxytoluene, 2-Carbethoxy-3,5-dihydroxytoluene, 4-Carbethoxy-5-methylresorcinol, 2,4-Dihydroxy-6-methylbenzoic Acid Ethyl Ester

C1316-1G C1316-5G
TCC1316-001GEA 93.22 CAD
TCC1316-001G TCC1316-005G
Ethyl-2,4-dihydroxy-6-methylbenzoate ≥98.0% (by titrimetric analysis)
Ethyl-2,4-dihydroxy-6-methylbenzoate

CAS Number: 2524-37-0
MDL Number: MFCD00010821
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 133


Other Product Names / Synonyms: 2-Carbethoxy-3,5-dihydroxytoluene, 4-Carbethoxy-5-methylresorcinol, 2,4-Dihydroxy-6-methylbenzoic Acid Ethyl Ester, 2-Ethoxycarbonyl-3,5-dihydroxytoluene, 4-Ethoxycarbonyl-5-methylresorcinol

Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
Melting Pt: 133 °C
MDL Number: MFCD00010821
CAS Number: 2524-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70a08cf6c11878aab3c43a5481025a891fee1e29&vid=70a08cf6c11878aab3c43a5481025a891fee1e29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR