3-Chloromethyl-5-nitrosalicylaldehyde ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Chloromethyl)-2-hydroxy-5-nitrobenzaldehyde

C1305-5G C1305-25G
TCC1305-005GEA 499.91 CAD
TCC1305-005G TCC1305-25G
3-Chloromethyl-5-nitrosalicylaldehyde ≥97.0% (by HPLC, titration analysis)
3-Chloromethyl-5-nitrosalicylaldehyde

CAS Number: 16644-30-7
MDL Number: MFCD00191331
Molecular Formula: C8H6ClNO4
Molecular Weight: 215.59
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 92

Formula: C₈H₆ClNO₄
MW: 215.5905 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00191331
CAS Number: 16644-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead36dd67d83027288aca151954902a55aaa2fc2&vid=ead36dd67d83027288aca151954902a55aaa2fc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR