1-Chloro-9,10-bis(phenylethynyl)anthracene ≥96.0% (by HPLC)
Supplier: TCI America
C1289-1G
C1289-10G
TCC1289-001GEA
72.68
CAD
TCC1289-001G
TCC1289-010G
1-Chloro-9,10-bis(phenylethynyl)anthracene ≥96.0% (by HPLC)
1-Chloro-9,10-bis(phenylethynyl)anthracene
CAS Number: 41105-35-5
MDL Number: MFCD00012047
Molecular Formula: C30H17Cl
Molecular Weight: 412.92
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 203
Formula:
C₃₀H₁₇Cl MW: 412.92 g/mol Melting Pt: 203 °C |
MDL Number:
MFCD00012047 CAS Number: 41105-35-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...