Triethyl orthochloroacetate ≥95.0%

Supplier: TCI America

C1288-1ML C1288-5ML
TCC1288-001MLEA 211.56 CAD
TCC1288-001ML TCC1288-005ML
Triethyl orthochloroacetate ≥95.0%
Triethyl orthochloroacetate

CAS Number: 51076-95-0
MDL Number: MFCD00142889
Molecular Formula: C8H17ClO3
Molecular Weight: 196.67
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2-Chloro-1,1,1-triethoxyethane, Orthochloroacetic Acid Triethyl Ester

Formula: C₈H₁₇ClO₃
Boiling Pt: 91 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00142889
CAS Number: 51076-95-0

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR