2-(4-Chlorobutoxy)tetrahydropyran ≥95.0%

Supplier: TCI America

C1279-5G C1279-25G C1279-1G
TCC1279-005GEA 190.07 CAD
TCC1279-005G TCC1279-25G 76219-670
2-(4-Chlorobutoxy)tetrahydropyran ≥95.0%
2-(4-Chlorobutoxy)tetrahydropyran

CAS Number: 41302-05-0
MDL Number: MFCD00142812
Molecular Formula: C9H17ClO2
Molecular Weight: 192.68
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Specific Gravity (20/20): 1.07

Formula: C₉H₁₇ClO₂
MW: 192.68 g/mol
Boiling Pt: 131 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00142812
CAS Number: 41302-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b30d6a2516c5c3de4fb9660520508bd72154afe8&vid=b30d6a2516c5c3de4fb9660520508bd72154afe8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR