4-Chloro-2-fluoro-1-nitrobenzene ≥96.0%

Supplier: TCI America

C1261-5G C1261-25G
TCC1261-005GEA 41.08 CAD
TCC1261-005G TCC1261-025G
4-Chloro-2-fluoro-1-nitrobenzene ≥96.0%
4-Chloro-2-fluoro-1-nitrobenzene

CAS Number: 700-37-8
MDL Number: MFCD00042211
Molecular Formula: C6H3ClFNO2
Molecular Weight: 175.54
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 48


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Chloro-2-fluoro-1-nitrobenzene

Formula: C₆H₃ClFNO₂
Melting Pt: 48 °C
MDL Number: MFCD00042211
CAS Number: 700-37-8
EINECS: 211-842-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a4e94c3abe1f93030c59667f0e0e474ae3c73f6&vid=afe550359e00971cad0d418bea419e03cadfc7df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR