2-Methylindole-3-carboxamide ≥98.0%

Supplier: TCI America

Synonyms: 3-Carbamoyl-2-methylindole

C1255-5G C1255-1G
TCC1255-5GEA 801.06 CAD
TCC1255-5G TCC1255-001G
2-Methylindole-3-carboxamide ≥98.0%
2-Methylindole-3-carboxamide

CAS Number: 67242-60-8
MDL Number: MFCD00143101
Molecular Formula: C10H10N2O
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 225


Other Product Names / Synonyms: 3-Carbamoyl-2-methylindole

Formula: C₁₀H₁₀N₂O
MW: 174.20 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00143101
CAS Number: 67242-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8821a29e822cae65dc801901d6eb3a23b9ce9b8&vid=b8821a29e822cae65dc801901d6eb3a23b9ce9b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR