4-Chloro-2,5-dimethoxynitrobenzene ≥98.0%

Supplier: TCI America

C1244-25G C1244-250G
TCC1244-025GEA 112.18 CAD
TCC1244-025G TCC1244-250G
4-Chloro-2,5-dimethoxynitrobenzene ≥98.0%
4-Chloro-2,5-dimethoxynitrobenzene

CAS Number: 6940-53-0
MDL Number: MFCD00024577
Molecular Formula: C8H8ClNO4
Molecular Weight: 217.61
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 141
Flash Point (°C): 77

Formula: C₈H₈ClNO₄
Melting Pt: 141 °C
Flash Pt: 77 °C
MDL Number: MFCD00024577
CAS Number: 6940-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95f88690554a54323a1f09cabdd8f9695bfe14f5&vid=95f88690554a54323a1f09cabdd8f9695bfe14f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR