4-Chloro-4'-hydroxybiphenyl ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4'-Chloro-[1,1'-biphenyl]-4-ol, 4'-Chlorobiphenyl-4-ol
C1239-1G
C1239-5G
TCC1239-001GEA
208.24
CAD
TCC1239-001G
TCC1239-5G
4-Chloro-4'-hydroxybiphenyl ≥98.0% (by titrimetric analysis)
4-Chloro-4'-hydroxybiphenyl
CAS Number: 28034-99-3
MDL Number: MFCD00143298
Molecular Formula: C12H9ClO
Molecular Weight: 204.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 146
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4'-Chlorobiphenyl-4-ol
Formula:
C₁₂H₉ClO MW: 204.65 g/mol Melting Pt: 146 °C |
MDL Number:
MFCD00143298 CAS Number: 28034-99-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...