3-(Cyanomethyl)-2,4,5-trimethylthiophene ≥98.0%
Supplier: TCI America
C1228-5G
C1228-1G
TCC1228-5GEA
446.35
CAD
TCC1228-5G
TCC1228-001G
3-(Cyanomethyl)-2,4,5-trimethylthiophene ≥98.0%
3-(Cyanomethyl)-2,4,5-trimethylthiophene
CAS Number: 112440-49-0
MDL Number: MFCD00143113
Molecular Formula: C9H11NS
Molecular Weight: 165.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 66
Formula:
C₉H₁₁NS Boiling Pt: 130 °C Melting Pt: 66 °C |
MDL Number:
MFCD00143113 CAS Number: 112440-49-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...