1-Chloro-2-(chloromethyl)-3,5-dioxahexane ≥95.0%
Supplier: TCI America
Synonyms:
1,3-Dichloro-2-(methoxymethoxy)propane, 1,1-Bis(chloromethyl)-2,4-dioxapentane
C1197-5G
C1197-25G
TCC1197-5GEA
71.1
CAD
TCC1197-5G
TCC1197-25G
1-Chloro-2-(chloromethyl)-3,5-dioxahexane ≥95.0%
1-Chloro-2-(chloromethyl)-3,5-dioxahexane
CAS Number: 70905-45-2
MDL Number: MFCD00130114
Molecular Formula: C5H10Cl2O2
Molecular Weight: 173.03
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Specific Gravity (20/20): 1.23
Other Product Names / Synonyms: 1,1-Bis(chloromethyl)-2,4-dioxapentane, 1,3-Dichloro-2-(methoxymethoxy)propane
Formula:
C₅H₁₀Cl₂O₂ MW: 173.03 g/mol Boiling Pt: 95 °C Density: 1.23 (20 °C) |
MDL Number:
MFCD00130114 CAS Number: 70905-45-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...