1-Chloro-2-iodoethane ≥97.0% stabilised for synthesis
Supplier: TCI America
C1190-5G
C1190-1G
TCC1190-005GEA
651.28
CAD
TCC1190-005G
TCC1190-001G
1-Chloro-2-iodoethane ≥97.0% stabilised for synthesis
1-Chloro-2-iodoethane
(stabilized with Copper chip)
CAS Number: 624-70-4
MDL Number: MFCD00041453
Molecular Formula: C2H4ClI
Molecular Weight: 190.41
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 2.11
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: 2-Chloroethyl Iodide, 2-Iodoethyl Chloride
Formula:
C₂H₄ClI Boiling Pt: 140 °C Melting Pt: –15.6 °C Density: 2.11 (20 °C) |
MDL Number:
MFCD00041453 CAS Number: 624-70-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...