4-Cyano-3-tetrahydrothiophenone ≥98.0%
Supplier: TCI America
C1164-25G
C1164-5G
TCC1164-25GEA
1047.54
CAD
TCC1164-25G
TCC1164-005G
4-Cyano-3-tetrahydrothiophenone ≥98.0%
4-Cyano-3-tetrahydrothiophenone
CAS Number: 16563-14-7
MDL Number: MFCD00059049
Molecular Formula: C5H5NOS
Molecular Weight: 127.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 71
Other Product Names / Synonyms: 4-Cyano-3-oxotetrahydrothiophene
Formula:
C₅H₅NOS Boiling Pt: 125 °C Melting Pt: 71 °C |
MDL Number:
MFCD00059049 CAS Number: 16563-14-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...